
Sodium hydroxide 5 mol/l (5 N) in aqueous solution, VOLUSOL® volumetric solution
Catalog # VWRS83947.290
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548