Order Entry
France
ContactUsLinkComponent
 
 
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
  FFCHFF1111000
 :  
 
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
  FFCHFF1111000
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :40 g/mol
 :∼105 °C (1013 hPa)
 :1.02 g/cm³ (25 °C)
 :Ambient
 :215-185-5
 :1824
 :8,III
 :GHS07
 :1310-73-2
 :NaOH
 :MFCD00003548