

Nitrilotriacetic acid ≥98%, white powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:139-13-9
:
Formula:C₆H₉NO₆
MDL Number:MFCD00004287
:191.14 g/mol
Merck Index:12,06675
Boiling Pt:498 °C (1013 hPa)
Melting Pt:242 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-355-7
UN:2923
ADR:8,II
:GHS08
:GHS07
Specification Test Results
- Assay:≥98%
- Clarity of solution:Passes
Specification Test Results
Frequently Bought Together