Toluene-3,4-dithiol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:496-74-2
:
Formula:C₇H₈S₂
MDL Number:MFCD00004844
:156.27
Merck Index:12,09669
Boiling Pt:185 °C (84 mmHg)
Melting Pt:27 °C
Density:1.18 g/cm³
Storage Temperature:Refrigerator
EINECS:207-828-3
UN:2810
:GHS07
Specification Test Results
- Appearance:White to yellow Low melting crystalline solid
- Infrared spectrum:Conforms
- GC:≥94 %
- HPLC:≥94 %
Specification Test Results
Frequently Bought Together