Order Entry
France
ContactUsLinkComponent
Sodium acetate 40% in aqueous solution
Sodium acetate 40% in aqueous solution
Catalog # PRLSMC307221.0500
Supplier:  PROLABO CMP
CAS Number:  
Sodium acetate 40% in aqueous solution
Catalog # PRLSMC307221.0500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :82.03 g/mol
Storage Temperature:Ambient
EINECS:204-823-8
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459