Sodium hydrogen carbonate 3.5 g/l in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Storage Temperature:Ambient
EINECS:205-633-8
Frequently Bought Together