Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium acetate
Sodium acetate 500 g/l in aqueous solution
Sodium acetate 500 g/l in aqueous solution
Sodium acetate 500 g/l in aqueous solution
PRLS85544.290
:
PROLABO CMP
:
Sodium acetate 500 g/l in aqueous solution
PRLS85544.290
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
82.03 g/mol
:
Ambient
:
204-823-8
:
127-09-3
:
H₃CCOONa
:
MFCD00012459