N-Methylethylamine
Catalog # MOLEM86123311
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:59.11 g/mol
Boiling Pt:36…37 °C (1013 hPa)
Density:0,6893 g/cm³ (20 °C)
EINECS:210-862-1
CAS Number:624-78-2
Formula:C₃H₉N
MDL Number:MFCD00009030