2-Hexynoic acid methyl ester
Catalog # MOLEM72661626
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:126.16 g/mol
EINECS:242-690-8
CAS Number:18937-79-6
Formula:C₇H₁₀O₂
MDL Number:MFCD00041642