Order Entry
France
ContactUsLinkComponent
Methyl crotonate
Methyl crotonate
Catalog # MOLE35484610-500G
Supplier:  Molekula
CAS Number:  
Methyl crotonate
Catalog # MOLE35484610-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :100.12 g/mol
Merck Index:13,02625
Boiling Pt:118 °C (1013 hPa)
Density:0.95 g/cm³ (20 °C)
EINECS:210-793-7
UN:3272
ADR:3,II
CAS Number:623-43-8
Formula:C₅H₈O₂
MDL Number:MFCD00009287