
3-Nitrophthalic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:641-70-3
:
Formula:C₈H₃NO₅
MDL Number:MFCD00005921
:193.12 g/mol
Melting Pt:159…162 °C
:M-Clarity™ quality level = MQ200
EINECS:211-373-6
:GHS07
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:159 - 162 °C
- Molar Mass:193.11 g/mol
Specification Test Results
Frequently Bought Together