Order Entry
France
ContactUsLinkComponent
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Catalog # 8.20248.1000
Supplier:  Merck
CAS Number:  
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Catalog # 8.20248.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    0.063 g/l (20 °C)
  • Molar Mass:
    146.23 g/mol
  • Boiling Point:
    110 - 115 °C
  • Vapor Pressure:
    1000 hPa (110 °C)
  • Flash Point:
    6 °C
  • Density:
    0.80 g/cm3 (20 °C)
  • Ignition Point:
    182 °C

Specification Test Results

About this item

 :
 :146.23 g/mol
Merck Index:12,03515
Boiling Pt:109 °C (1013 hPa)
Melting Pt:< –25 °C
Density:0.798 g/cm³ (20 °C)
Flash Pt:12 °C
EINECS:203-733-6
UN:3107
ADR:5.2,
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS08
Storage Temperature:Ambient
CAS Number:110-05-4
Formula:C₈H₁₈O₂
MDL Number:MFCD00008803