Aniline hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride
CAS Number:142-04-1
:
Formula:C₆H₈ClN
MDL Number:MFCD00012958
Boiling Pt:245
Melting Pt:198
Density:1.2215 g/cm³ (20 °C)
Molecular Weight:129.59 g/mol
Flash Pt:193
EINECS:205-519-8
UN:1548
ADR:6.1 III
Merck Index:13,00616
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Hill Formula:C₆H₈ClN
- Chemical formula:C₆H₅NH₂ * HCl
- Molar Mass:129.58 g/mol
- Boiling point:245 °C (1013 hPa)
- Flash point:193 °C
- Melting Point:199 - 202 °C
- Bulk density:500 kg/m³
- Solubility:1070 g/l (25 °C)
- LD 50 oral:LD50 Rat 840 mg/kg
- Storage class:6.1A Combustible substances, toxic
- WGK:WGK 2 water endangering
- R Phrase:R 23/24/25-40-41-43-48/23/24/25-68-50
- S Phrase:S 26-27-36/37/39-45-61-63
Specification Test Results
Frequently Bought Together