
Crotonic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-93-7
:3724-65-0
Formula:C₄H₆O₂
MDL Number:MFCD00002701
:86.09 g/mol
Merck Index:13,02625
Boiling Pt:185…199 °C (1013 hPa)
Melting Pt:71…72 °C
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-533-9
UN:2823
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:90 °C
:GHS06
Specification Test Results
- Solubility:6.2 g/l (20 °C)
- Melting Point:70 - 73 °C
- Molar Mass:86.09 g/mol
- Bulk Density:543 kg/m3
- Boiling Point:185.0 - 199.5 °C (1013 hPa)
- Vapor Pressure:0.25 hPa (20 °C)
- Flash Point:90 °C
- Density:1.02 g/cm3 (20 °C)
- pH:3 (10 g/l, H2O, 20 °C)
- Ignition Point:500 °C
Specification Test Results
Frequently Bought Together