Order Entry
France
ContactUsLinkComponent
 
Corrosive
Health
Environment
Tetrachlorophthalic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.00741.0100
Supplier:  Merck
CAS Number:  
Corrosive
Health
Environment
Tetrachlorophthalic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.00741.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    253 - 256 °C
  • Molar Mass:
    285.89 g/mol
  • Boiling Point:
    350 °C (1.013 hPa)
  • Vapor Pressure:
    0.0021 kPa (145 °C)

Specification Test Results

About this item

 :
 :285.9 g/mol
Boiling Pt:371 °C (1013 hPa)
Density:1.52 g/cm³ (275 °C)
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:204-171-4
UN:3077
 :GHS05
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:117-08-8
Formula:C₈Cl₄O₃
MDL Number:MFCD00005920