
Sodium nitroprusside dihydrate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
:13755-38-9
:
:Na₂[Fe(CN)₅NO]·2H₂O
:MFCD00149192
:13,08722
:1.72 g/cm³ (20 °C)
:Ambient
:6.1,II
:M-Clarity™ quality level = MQ100
:297.95 g/mol
:238-373-9
:1588
:GHS06
- Solubility:400 g/l (20 °C) (slow decomposition)
- Molar Mass:297.95 g/mol
- Bulk Density:1000 kg/m3
- Density:1.71 g/cm3 (20 °C)
- pH:5 (50 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Frequently Bought Together