Potassium iodide 40 g/l in aqueous solution
Catalog # FFCHFF22675000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405