Order Entry
France
ContactUsLinkComponent
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF0281000
Supplier:  FF CHEMICALS
CAS Number:  
EDTA disodium salt 0.05 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF0281000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672