Organic reference standard, 2,2-Dichloropropane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12481200ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:594-20-7
- Description:2,2-Dichloropropane 100 µg/ml in methanol
- Formula:C₃H₆Cl
- Description:2,2-Dichloropropane