1-(4-Methylphenyl)-2-thiourea
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 4-Methylphenylthiourea, N-(p-Tolyl)thiourea, N-(4-Methylphenyl)thiourea, p-Tolylthiourea, 1-(p-Tolyl)thiourea, 1-(4-Tolyl)thiourea
CAS Number:622-52-6
:
Formula:(C₆H₄(CH₃))NC(S)NH₂
MDL Number:MFCD00041190
:166.25 g/mol
Boiling Pt:295 °C (1013 hPa)
Melting Pt:195…196 °C
EINECS:210-740-8
UN:2811
ADR:6.1,III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together