
Sodium nitroprusside dihydrate, Millipore®
Catalog # 567538-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1588
ADR:6.1,III
:297.95 g/mol
EINECS:238-373-9
:GHS06
CAS Number:13755-38-9
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192