
Benzyl bromoacetate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5437-45-6
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00000190
:229.07 g/mol
Boiling Pt:166…170 °C (22 mmHg)
Density:1.44 g/cm³
Flash Pt:>109 °C
EINECS:226-611-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5430 to 1.5450 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
Frequently Bought Together