Order Entry
France
ContactUsLinkComponent
O-Phenetidine 99%
Toxic
Health
O-Phenetidine 99%
Certificates
undefined
Toxic
Health
O-Phenetidine 99%
Certificates

Specification Test Results

  • Appearance:
    Light yellow to red Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5540 to 1.5560 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together