Allylthiourea 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
CAS Number:109-57-9
:
Formula:H₂CCHCH₂NHC(S)NH₂
MDL Number:MFCD00004940
:116.19 g/mol
Merck Index:12,09500
Boiling Pt:191 °C (1013 hPa)
Melting Pt:70…78 °C
Density:1,219 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-683-5
UN:2811
ADR:6.1,III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light beige Powder or crystals or granules and/or chunks
- Infrared spectrum:Conforms
- Melting point:70 to 78 °C
- Loss on drying:≤1.5 % (50 °C, 2 hrs)
- Titration with HClO4:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together