
trans-Cinnamaldehyde 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14371-10-9
:
Formula:C₉H₈O
MDL Number:MFCD00007000
:132.16 g/mol
Merck Index:12,02319
Boiling Pt:250…252 °C (760 mmHg)
Melting Pt:–7.5 °C
Density:1.05 g/cm³
Flash Pt:71 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6210 to 1.6230 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:2,6-di-tert.-Butyl-p-cresol
Specification Test Results
Frequently Bought Together