Order Entry
France
ContactUsLinkComponent
Methyl 5-(difluoromethyl)nicotinate 95%
Irritant
Methyl 5-(difluoromethyl)nicotinate 95%
Catalog # APOSPC55854-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 5-(difluoromethyl)nicotinate 95%
Catalog # APOSPC55854-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.14 g/mol
 :GHS07
CAS Number:1550967-83-3
Formula:C₈H₇F₂NO₂
MDL Number:MFCD25476641