Order Entry
France
ContactUsLinkComponent
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)
Catalog # AATB21487
Supplier:  AAT Bioquest
CAS Number:  
undefined
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)
Catalog # AATB21487
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :607.51 g/mol
CAS Number:162112-35-8
Formula:C₃₀H₄₅Br₂N₃
MDL Number:MFCD00467819