Order Entry
France
ContactUsLinkComponent
OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
Irritant
OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
Catalog # AATB20506
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
Catalog # AATB20506
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :1114.28 g/mol
ADR:5,II
 :GHS07