Order Entry
France
ContactUsLinkComponent
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 20 mM in water ≥95%
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 20 mM in water ≥95%
Catalog # AATB17525
Supplier:  AAT Bioquest
CAS Number:  
undefined
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 20 mM in water ≥95%
Catalog # AATB17525
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :533.88 g/mol (anhy)
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl·xH₂O
MDL Number:MFCD00012679