Order Entry
France
ContactUsLinkComponent
 
Native Streptavidin (from Streptomyces avidinii), Alexa Fluor® 405
 
undefined
Native Streptavidin (from Streptomyces avidinii), Alexa Fluor® 405

 

  • Conjugation:
    Alexa Fluor® 405
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin| Streptomyces avidinii

 

Frequently Bought Together