Order Entry
France
ContactUsLinkComponent
ReadiLink™Rapid mFluor™ Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
ReadiLink™Rapid mFluor™ Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1131
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™Rapid mFluor™ Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1131
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid mFluor™ red 780 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
mFluor™ dyes are developed for flow cytometry-focused applications.
  • Red-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.