Order Entry
France
ContactUsLinkComponent
2,3-Butanediol for synthesis, Sigma-Aldrich®
2,3-Butanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,3-Butanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1000 g/l (20 °C)
  • Melting Point:
    18 - 20 °C
  • Molar Mass:
    90.12 g/mol
  • Boiling Point:
    180 - 182 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    96 °C DIN 51758
  • Refractive Index:
    1.4348 (20 °C, 589 nm)
  • Explosion Limit:
    3.1 - 11.4 %(V)
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    7 (H2O, 20 °C) Aqueous solution
  • Ignition Point:
    395 °C DIN 51794

Specification Test Results

Related Information

Frequently Bought Together