Order Entry
France
ContactUsLinkComponent
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
Catalog # AATB21480
Supplier:  AAT Bioquest
CAS Number:  
undefined
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
Catalog # AATB21480
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
Formula:C₂₉H₄₀N₂O₃S