Order Entry
France
ContactUsLinkComponent
Cell Meter™ Live Cell ATP Assay Kit
Cell Meter™ Live Cell ATP Assay Kit
Catalog # AATB23015
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Live Cell ATP Assay Kit
Catalog # AATB23015
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ Live cell ATP assay kit
  • Tests per kit:
    100 Tests

Specifications

About this item

Adenosine triphosphate (ATP) plays a fundamental role in cellular energetics, metabolic regulation and cellular signaling.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.