Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 750)
Supplier: AAT Bioquest
Certificates
About this item
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
- Bright signal
- Increased photostability
- Minimal quenching on proteins
Caution: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Specifications
- Catalog No:
- AATB16842
- AATB16720
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- iFluor® 750
- iFluor® 750
- Clone:
- IgG
- IgG
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Cross adsorption:
- Min×Human
- Min×Human
Specifications
Frequently Bought Together