
Phalloidin Amine for biomolecular labeling
Catalog # AATB5302
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 µG
- Seq:1286
Specifications
About this item
:
:901.91 g/mol
ADR:5,II
:GHS07
CAS Number:87876-22-0