Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FMOC-Lys(TQ3)-OH
FMOC-Lys(TQ3)-OH for biomolecular labeling
FMOC-Lys(TQ3)-OH for biomolecular labeling
Catalog # AATB5258
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
FMOC-Lys(TQ3)-OH for biomolecular labeling
Catalog # AATB5258
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
Environmentally Preferable:
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details