0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
FMOC-Asp(TQ2)-OH
FMOC-Asp(TQ2)-OH for biomolecular labeling
FMOC-Asp(TQ2)-OH for biomolecular labeling
Catalog # AATB5206
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
FMOC-Asp(TQ2)-OH for biomolecular labeling
Catalog # AATB5206
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
Environmentally Preferable:
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details