Fmoc-Sta-OH ≥95%
Supplier: AAT Bioquest
Jump to:
About this item
(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.
Synonyms: (3S,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-6-methylheptanoic acid
- CAS Number:158257-40-0
- Formula:C₂₃H₂₇NO₅
- MDL Number:MFCD00065675
- Molecular Weight:397.48 g/mol
- ADR:5,II
2 Options Available Below
Product Details & Documents
(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.
Synonyms: (3S,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-6-methylheptanoic acid
- CAS Number:158257-40-0
- Formula:C₂₃H₂₇NO₅
- MDL Number:MFCD00065675
- Molecular Weight:397.48 g/mol
- ADR:5,II



