Anti-MAP2K1|MAP2K2 Rabbit Recombinant Antibody [clone: MEK12S221-D3]
ABNORAB02830.200UL
: Abnova
- Antigen symbol:MAP2K1|MAP2K2
- Clonality:Recombinant
- Clone:MEK12S221-D3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human MAP2K1|MAP2K2.
- Pk:200 µl