Order Entry
France
ContactUsLinkComponent
Anti-P-glycoprotein Rabbit Recombinant Antibody [clone: MRK16]
Anti-P-glycoprotein Rabbit Recombinant Antibody [clone: MRK16]
Catalog # ABNORAB03755.200UG
Supplier:  Abnova
Anti-P-glycoprotein Rabbit Recombinant Antibody [clone: MRK16]
Catalog # ABNORAB03755.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    P-glycoprotein
  • Clonality:
    Recombinant
  • Clone:
    MRK16
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human P-glycoprotein.
  • Pk:
    200 µG

Specifications

About this item