Anti-GLB1 Rabbit Recombinant Antibody [clone: 13R4]
Catalog # ABNORAB03692.200UG
Supplier: Abnova
Specifications
- Antigen symbol:GLB1
- Clonality:Recombinant
- Clone:13R4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GLB1.
- Pk:200 µG