Anti-IRF3 Rabbit Recombinant Antibody [clone: R02-6B0]
ABNORAB01424.100UL
: Abnova
- Antigen symbol:IRF3
- Clonality:Recombinant
- Clone:R02-6B0
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human IRF3.
- Pk:100 µl