Order Entry
France
ContactUsLinkComponent
Anti-IQGAP2 Rabbit Recombinant Antibody [clone: R08-2C6]
Anti-IQGAP2 Rabbit Recombinant Antibody [clone: R08-2C6]
Catalog # ABNORAB01425.100UL
Supplier:  Abnova
Anti-IQGAP2 Rabbit Recombinant Antibody [clone: R08-2C6]
Catalog # ABNORAB01425.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    IQGAP2
  • Clonality:
    Recombinant
  • Clone:
    R08-2C6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human IQGAP2.
  • Pk:
    100 µl

Specifications

About this item