Anti-HCK Rabbit Recombinant Antibody [clone: R02-5C1]
ABNORAB01438.100UL
: Abnova
- Antigen symbol:HCK
- Clonality:Recombinant
- Clone:R02-5C1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HCK.
- Pk:100 µl