Anti-OPTN Rabbit Recombinant Antibody [clone: R01-2C5]
Catalog # ABNORAB01394.100UL
Supplier: Abnova
Specifications
- Antigen symbol:OPTN
- Clonality:Recombinant
- Clone:R01-2C5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human OPTN.
- Pk:100 µl