Anti-NDUFS2 Rabbit Recombinant Antibody [clone: R08-4D7]
ABNORAB01405.100UL
: Abnova
- Antigen symbol:NDUFS2
- Clonality:Recombinant
- Clone:R08-4D7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NDUFS2.
- Pk:100 µl