Order Entry
France
ContactUsLinkComponent
 
Anti-MIF Rabbit Recombinant Antibody [clone: R05-8H5]
  ABNORAB02105.100UL
 :  Abnova
Anti-MIF Rabbit Recombinant Antibody [clone: R05-8H5]
  ABNORAB02105.100UL
 :  Abnova

 

  • Antigen symbol:
    MIF
  • Clonality:
    Recombinant
  • Clone:
    R05-8H5
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human MIF.
  • Pk:
    100 µl