Anti-ISL1 Rabbit Recombinant Antibody [clone: R05-2C3]
Catalog # ABNORAB02159.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ISL1
- Clonality:Recombinant
- Clone:R05-2C3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ISL1.
- Pk:100 µl