Cerex® MAESTRO SPE Column
TCAT613-0151R
: Tecan
- Particle Size:10 µm
- Capacity:1 ml
- Description:Column Cerex MAESTRO
- Bed weight:15 mg
- Phase:MAESTRO
- Pk:96
Silica-based sorbent with C18 bonding in proprietary coverage.
- Reversed-phased
- Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.